Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ceramic Discs, Round, AL23, with Hole, Thermo Scientific
Supplier: Thermo Scientific
33143-KF 33146-KF 33144-KF 33142-KF 33140-KF 33146-KF 033147-KF 33142-KF 33144-KF 33147-KF 033140-KF 33143-KF
AA33144-KFEA 48.02 USD InStock
CAAA33146-KF CAAA33147-KF CAAA33144-KF AA33144-KF AA33146-KF AA33147-KF CAAA33142-KF CAAA33143-KF AA33142-KF AA33143-KF CAAA33140-KF AA33140-KF
Ceramic Discs, Round, AL23, with Hole, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al2O3 Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty