Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Synonyms: (R,R)-(+)-Potassium sodium tartrate tetrahydrate (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate L(+)-Tartaric acid potassium sodium salt tetrahydrate (+)-Potassium sodium L-tartrate tetrahydrate Potassium sodium L(+)-tartrate tetrahydrate

33241-A4 033241-22 33241-22 33241-36 33241-A4 033241-36
AA33241-A4EA 401.99 USD InStock
AA33241-A4 AA33241-36 CAAA33241-22 CAAA33241-36 CAAA33241-A4 AA33241-22
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% ACS
L(+)-Potassium sodium tartrate tetrahydrate


Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: 1.77
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg
Order Now
Specification Test Results
Assay (unspecified) 99.0-102.0% C₄H₄KNaO₆.4H₂O
Impurity content Ca 0.005%
Insoluble material 0.005%
pH of a 5% solution 6.0-8.5 at 25°C
Impurity content Chloride (as Cl) 0.0015
Impurity content Fe 0.001%
Impurity content PO₄ 0.002%
Impurity content SO₄ 0.005%
Impurity content NH₄ 0.002%
Trace Metals (as Pb) 5ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty