Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 68.0 - 70.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aqua fortisAzotic acidHydrogen nitrate

033260-P7 033260-P2 33260-R1
AA33260-P2EA 73.45 USD InStock
AA33260-P2 AA-33260-R1 AA33260-P7
Nitric acid 68.0 - 70.0% ACS
Nitric acid


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 121 °C
Melting Pt: –42 °C
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608
Order Now
Specification Test Results
Color (APHA) 10
Residue after ignition 5 ppm
Cl 0.5 ppm
SO3 1 ppm
As 0.01 ppm
Heavy Metals (as Pb) 0.2 ppm
Fe 0.2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty