Synonyms:
Ethanedioic acid dihydrate
033262-A1
33262-A7
33262-A1
033262-A7
33262-18
033262-18
33262-30
033262-30
AA33262-18EA
61.61
USD
InStock
CAAA33262-A7
AA33262-18
CAAA33262-18
AA33262-A7
CAAA33262-30
AA33262-A1
AA33262-30
CAAA33262-A1
Oxalic acid dihydrate 99.5-102.5% ACS
Oxalic acid dihydrate
As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation