Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid dihydrate 99.5-102.5% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethanedioic acid dihydrate

033262-A1 33262-A7 33262-A1 033262-A7 33262-18 033262-18 33262-30 033262-30
AA33262-18EA 61.61 USD InStock
CAAA33262-A7 AA33262-18 CAAA33262-18 AA33262-A7 CAAA33262-30 AA33262-A1 AA33262-30 CAAA33262-A1
Oxalic acid dihydrate 99.5-102.5% ACS
Oxalic acid dihydrate
As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation


Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101…105 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg
Order Now
Specification Test Results
Insoluble matter 0.005
Residue after ignition 0.01%
Cl 0.002%
SO3 0.005%
Ca 0.001%
Nitrogen compounds (as N) 0.001%
Heavy Metals (as Pb) 5 ppm
Fe 2 ppm
Substances darkened by hot sulfuric acid Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty