Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orthophosphoric acid ≥85% (w/w) in aqueous solution ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: o-Phosphoric acidPhosphoric acidPhosphoric acid ortho

33266-D6 033266-A1 033266-22 33266-22 033266-D6 33266-A1
AA33266-D6EA 160.22 USD InStock
CAAA33266-D6 AA33266-D6 CAAA33266-22 CAAA33266-A1 AA33266-A1 AA33266-22
Orthophosphoric acid ≥85% (w/w) in aqueous solution ACS
Orthophosphoric acid


Formula: H₃PO₄
MW: 98 g/mol
Melting Pt: 19…21 °C
Density: 1.70
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Specification Test Results
Color (APHA) 10
Insoluble matter 0.001%
Cl 3 ppm
NO3 5 ppm
SO3 0.003%
Volatile acids (as CH3COOH) 0.001%
Sb 0.002%
Ca 0.002%
Mg 0.002%
As 1ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty