Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicic acid hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

033270-22 33270-22 033270-36 33270-36
AA33270-36EA 312.07 USD InStock
AA33270-36 CAAA33270-36 AA33270-22 CAAA33270-22
Silicic acid hydrate, Reagent Grade
Silicic acid hydrate
Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.


Formula: SiO₂.xH₂O
MW: 60.09 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS Number: 1343-98-2
EINECS: 215-683-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty