033270-22
33270-22
033270-36
33270-36
AA33270-36EA
312.07
USD
InStock
AA33270-36
CAAA33270-36
AA33270-22
CAAA33270-22
Silicic acid hydrate, Reagent Grade
Silicic acid hydrate
Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.