Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfide 20 - 24% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

33286-AP 33286-AE 033286-AE 033286-AP
AA33286-AEEA 144.99 USD InStock
AA33286-AE CAAA33286-AP AA33286-AP CAAA33286-AE
Ammonium sulfide 20 - 24% in aqueous solution
Ammonium sulphide


Formula: (NH₄)₂S
MW: 68.14 g/mol
Density: 0,997
Storage Temperature: Ambient
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty