Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium fluoride 97%, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning

033297-22 33297-36 33297-22 033297-36
AA33297-22EA 26.59 USD InStock
CAAA33297-36 AA33297-22 CAAA33297-22 AA33297-36
Calcium fluoride 97%, REacton®, Reagent Grade
Calcium fluoride
Spectroscopy, electronics, laser, high temperature dry-film lubricants


Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: ∼2500 °C
Melting Pt: 1403 °C
Density: 3.18
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty