Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 37% in aqueous solution stabilized ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Formalin

Stabilization: Stabilized with methanol 10-15%

33314-K2 33314-K7 33314-M1 033314-K7 033314-K2 033314-M6 033314-AP 033314-M1 33314-M6 33314-AP
AA33314-M6EA 395.46 USD InStock
CAAA33314-K7 AA33314-M6 AA33314-K7 CAAA33314-AP AA33314-AP CAAA33314-M1 AA33314-M1 CAAA33314-K2 AA33314-K2 CAAA33314-M6
Formaldehyde 37% in aqueous solution stabilized ACS
Formaldehyde


Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 97 °C
Melting Pt: –15 °C
Density: 1.083
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 3,III
Order Now
Specification Test Results
Color (APHA) 10
Residue after ignition 0.005% max.
Titratable acid 0.006meq/g max.
Cl 5 ppm max.
SO3 0.002% max.
Heavy Metals (as Pb) 5 ppm max.
Fe 5 ppm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty