Synonyms:
Lead monoxide
33330-36
033330-36
33330-A3
033330-A3
AA33330-A3EA
84.18
USD
InStock
AA33330-A3
CAAA33330-36
CAAA33330-A3
AA33330-36
Lead(II) oxide low silver
Lead (II) oxide
As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores