Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium hydrogen orthophosphate trihydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Synonyms: Potassium phosphate dibasic trihydrate di-Potassium hydrogen orthophosphate trihydrate

33365-30 033365-A1 033365-30 33365-A1
AA33365-30EA 93.42 USD InStock
AA33365-30 CAAA33365-A1 AA33365-A1 CAAA33365-30
Dipotassium hydrogen orthophosphate trihydrate, REacton®, Reagent Grade
di-Potassium hydrogen orthophosphate trihydrate


Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty