Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogenarsenate heptahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium arsenate dibasic heptahydrate

033373-14 33373-14 33373-22 033373-22
AA33373-14EA 223.21 USD InStock
AA33373-14 CAAA33373-22 CAAA33373-14 AA33373-22
Disodium hydrogenarsenate heptahydrate 98.0-102.0% ACS
Disodium hydrogenarsenate heptahydrate


Formula: HAsNa₂O₄·7H₂O
MW: 312.01 g/mol
Melting Pt: 130 °C (dec.)
Density: 1.88
Storage Temperature: Ambient
MDL Number: MFCD00149179
CAS Number: 10048-95-0
EINECS: 231-902-4
UN: 1685
ADR: 6.1,II
Order Now
Specification Test Results
Insoluble matter 0.005%
As2O3 0.01%
Cl 0.001%
NO3 0.005%
SO3 0.01%
Heavy Metals (as Pb) 0.002%
Fe 0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty