Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trilithium nitride ≥99.4% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Lithium nitride Trilithium nitride

35479-09 035479-09 35479-18 035479-18
AA35479-18EA 295.99 USD InStock
CAAA35479-18 CAAA35479-09 AA35479-18 AA35479-09
Trilithium nitride ≥99.4% (metals basis)
tri-Lithium nitride


Formula: Li₃N
Melting Pt: 845 °C
Density: 1.30
Storage Temperature: Ambient
MDL Number: MFCD00016186
CAS Number: 26134-62-3
EINECS: 247-475-2
UN: 2806
ADR: 4.3,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty