Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Phosphate, Ion Chromatography Standard Solution, Specpure®, PO4-3 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
035548.AP 35548-AE 035548-AE 35548-AP
AAA35548-APEA 234.34 USD InStock
CAAAA35548-AP AAA35548-AP AA35548-AE CAAA35548-AE
Phosphate, Ion Chromatography Standard Solution, Specpure®, PO4-3 1000 µg/ml, Thermo Scientific
Standards Anionic Standards Phosphate Standards
Keep refrigerated

Molecular Formula: KH2PO4 in H2O
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00145108

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty