Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid standardised solution 1.0 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

35628-AP 035628-AP
AA35628-APEA 115 USD InStock
CAAA35628-AP AA35628-AP
Lactic acid standardised solution 1.0 N in aqueous solution
Lactic acid


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty