Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(IV) sulfate standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cerium disulphateCeric sulphateCerium disulfateCeric sulfateCerium(IV) sulfateCerium(IV) sulphate

35650-K2 035650-K7 035650-K2 35650-K7
AA35650-K2EA 156.94 USD InStock
CAAA35650-K2 AA35650-K2 AA35650-K7 CAAA35650-K7
Cerium(IV) sulfate standardised solution 0.1 N in aqueous solution
Cerium (IV) sulphate


Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8,III
Merck Index: 13,02002
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty