Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc chloride, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Butter of zincZinc (II) chloride

35726-06 35726-22 35726-14 35726-22 035726-06 035726-14
AA35726-22EA 571.99 USD InStock
CAAA35726-22 AA35726-22 CAAA35726-14 AA35726-14 CAAA35726-06 AA35726-06
Zinc chloride, ultra dry ≥99.999% (metals basis)
Zinc chloride


Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty