Would you like to log in again?
High Form Nickel Crucibles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
035967-KT 036040.KT 035904-KT 035883-KT 035925-KT 035946-KT
AA36040-KTEA 203.91 USD InStock
AA36040-KT AA35925-KT AA35904-KT AA35946-KT AA35967-KT AA35883-KT
High Form Nickel Crucibles, Thermo Scientific
Crucibles
Nickel crucibles offer high resistance to dilute alkalies at a very low cost per crucible.
  • Life expectancy of a nickel crucible is from 4 to 6 fusions
  • Nickel is completely resistant to phosphoric acid and highly resistant to the corrosive effect of strong alkalies
  • No dry gases are actively corrosive to nickel at atmospheric temperature
  • Nickel is also resistant to dry hydrogen chloride, hydrogen fluoride, and chlorine up to about 535°C
  • Nickel is not affected by steam at temperatures usually encountered
In choosing crucibles for laboratory work, nickel can be effective with regard to cost per crucible, and for use in fusions where zirconium or other metals cannot be used.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty