Would you like to log in again?
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
036083.KT 035968-KT 35968-KT 035947-KT 35926-KT 36083-KT 036039.KT 035926-KT 35997-KT 036058.KT 35947-KT 035905-KT 36058-KT 36039-KT
AA36039-KTEA 248.32 USD InStock
CAAA35926-KT AA36039-KT CAAA35968-KT AA36083-KT CAAA36058-KT AA35997-KT CAAA35947-KT AA35926-KT CAAA36039-KT AA35905-KT AA36058-KT AA35947-KT CAAA36083-KT AA35968-KT
Low Form Zirconium Crucibles, Thermo Scientific
Crucibles
Zirconium crucibles are handmade to an exacting tolerance for uniform wall thickness
  • Special apparatus is not required for handling hot zirconium crucibles
  • Sudden contact with cold, metallic surfaces will have no deleterious effect on a zirconium crucible
  • Zirconium crucibles require a minimum of specialized care; smoothing and shaping is not a special consideration
  • The inherent strength of zirconium precludes the necessity of reinforced rims and thicker bottoms
These crucibles are produced under the most stringent requirements to ensure ultimate tensile strength, yield strength, elongation, and chemical purity.
Only high-purity zirconium material is used.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty