Would you like to log in again?
Low Form Inconel® Alloy 601 Crucibles, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
035928-KT 035994-KT 036037.KT 35970-KT 036016.KT 036056.KT 036085.KT 035886-KT
AA35928-KTEA 90.24 USD InStock
AA35928-KT AA36016-KT AA36085-KT AA36037-KT AA36056-KT AA35970-KT AA35886-KT AA35994-KT
Low Form Inconel® Alloy 601 Crucibles, Thermo Scientific
Crucibles
Inconel® nickel-chromium-iron alloy 601 is a general-purpose engineering material for applications that require resistance to heat and corrosion
  • Ideal for high-temperature applications requiring resistance to oxidation and spalling
  • The alloy is unaffected by rapid changes from hot to cold
  • Retains mechanical strength at elevated temperatures
The high resistance of Inconel® Alloy 601 to oxidation, carburization, or sulfidation make it well suited for vessels used in determining moisture, volatiles, fixed-carbon, and ash located in most coal and coke products, or wood pulp or fiber. It is also recommended for use in drying and ashing biological materials whose residues are soluble in dilute acid or alkali for subsequent analysis. Trace-level determinations of principal constituent elements are excluded. The vessels can be cleaned simply by scouring with sea-sand or some other mild abrasive.
It offers excellent resistance to oxidation in the 1000 to 1200°C temperature range, and provides good corrosion resistance to many acid and aqueous salt solutions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty