Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

036187-03 36187-06 36187-03 036187-06
AA36187-06EA 1841.99 USD InStock
AA36187-06 CAAA36187-06 AA36187-03 CAAA36187-03
Sodium selenide ≥99.8% (metals basis)
Sodium selenide


Formula: Na₂Se
MW: 124.94 g/mol
Melting Pt: >875 °C
Density: 2,625
Storage Temperature: Ambient
MDL Number: MFCD00014240
CAS Number: 1313-85-5
EINECS: 215-212-0
UN: 3283
ADR: 6.1,II
Merck Index: 12,08821
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty