Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Resorcinol 99.0-100.5% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,3-Benzenediol 1,3-Dihydroxybenzene

36248-22 36248-14 36248.14 36248-36 036248-36 036248-22
AA36248-22EA 93.98 USD InStock
CAAA36248-36 AA36248-22 CAAAA36248-14 AAA36248-14 CAAA36248-22 AA36248-36
Resorcinol 99.0-100.5% ACS
Resorcinol


Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 280…281 °C
Melting Pt: 109…112 °C
Density: 1.272
Flash Pt: 171 °C (339 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1,III
Merck Index: 12,08323
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/84/2299384.jpg
Order Now
Specification Test Results
Form Crystalline
Insoluble matter 0.005%
Residue after ignition 0.01%
Titratable acid 0.004meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty