Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Synonyms: Ytterbium oxide Diytterbium trioxide

36252-14 036252-14 36252-22 036252-22
AA36252-14EA 184.99 USD InStock
AA36252-14 CAAA36252-14 AA36252-22 CAAA36252-22
Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis)
Ytterbium (III) oxide


Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2227 °C
Density: 9.2
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0
Merck Index: 14,10105
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty