Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury (II) oxide, yellow ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Mercury monoxide yellow

36261-22 036261-14 036261-22
AA36261-14EA 58.73 USD InStock
AA36261-14 CAAA36261-22 AA36261-22
Mercury (II) oxide, yellow ≥99.0% ACS
Mercury (II) oxide


Formula: HgO
MW: 216.59 g/mol
Melting Pt: 500 °C (dec.)
Density: 11.14
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 13,05908
Order Now
Specification Test Results
Insoluble in dilute hydrochloric acid 0.03%
Residue after reduction 0.05%
Cl 0.025%
SO3 0.01%
Nitrogen compounds (as N) 0.005%
Fe 0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty