Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) sulfate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Mercuric sulphate Mercury sulphate

36286-36 036286-14 36286-14 036286-22 036286-36
AA36286-14EA 37.99 USD InStock
AA36286-14 AA36286-36 CAAA36286-36 CAAA36286-14 AA36286-22
Mercury(II) sulfate ≥98.0% ACS
Mercury (II) sulphate
Applications: Primary battery electrolyte, catalyst in conversion of acetylene to acetaldehyde


Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: Dec.
Density: 6.470
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1,II
Merck Index: 13,05914
Order Now
Specification Test Results
Residue after reduction 0.02%
Cl 0.003%
NO7 Pass test
Fe 0.005%
Mercurous mercury (as Hg) 0.15%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty