Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid glacial ≥99.7% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethanoic acidGlacial acetic acidMethanecarboxylic acidVinegar acid

36289-AE 36289-AP 036289-AY 36289-K3 036289-K3 036289-AP 036289-AE 36289-AY
AA36289-AYEA 160.92 USD InStock
AA36289-AY AA36289-K3 AA36289-AE CAAA36289-K3 CAAA36289-AY AA36289-AP CAAA36289-AE CAAA36289-AP
Acetic acid glacial ≥99.7% ACS
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118.1 °C
Melting Pt: 16.6 °C
Density: 1.049
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Color (APHA) 10
Dilution Test Pass test
Evaporation residue 0.001%
(CH3CO)2O 0.01%
Cl 1 ppm
SO3 0.1%
Heavy Metals (as Pb) 0.5 ppm
Fe 0.2 ppm
Substances reducing dichromate Pass test
Substances reducing permanganate P.T. Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty