Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azelaic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,7-Heptanedicarboxylic acid 1,9-Nonanedioic acid

036308-59 036308-22 036308-14 36308-22 36308-14 36308-59
AA36308-14EA 41.03 USD InStock
AA36308-14 CAAA36308-59 AA36308-59 CAAA36308-14 CAAA36308-22 AA36308-22
Azelaic acid 98%
Azelaic acid


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 286 °C (100 mmHg)
Melting Pt: ∼ 102 °C
Density: 1,225
Flash Pt: 215 °C (419 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty