Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-oneD-Isoascorbic acid

36366.22 36366-36 36366-22 36366.36
AA36366-22EA 58.68 USD InStock
CAAA36366-22 CAAA36366-36 AA36366-22 AA36366-36
D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)
D(-)-Isoascorbic acid


Formula: C₆H₈O₆
MW: 176.12 g/mol
Melting Pt: 169…172 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005378
CAS Number: 89-65-6
EINECS: 201-928-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty