Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Tartaric acid ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R,R)-(+)-Tartaric acid natural tartaric acid (R,R)-(+)-2,3-Dihydroxysuccinic acid

36405-36 36405-A3 36405-22 036405-36 036405-22 036405-A3
AA36405-36EA 101.32 USD InStock
AA36405-36 AA36405-A3 CAAA36405-36 CAAA36405-A3 AA36405-22 CAAA36405-22
L(+)-Tartaric acid ACS
L(+)-Tartaric acid
Useful as a resolving agent


Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 168…170 °C
Density: 1.760
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/32/2297232.jpg
Order Now
Specification Test Results
Insoluble matter 0.005
Residue after ignition 0.02
Cl 0.001
C2O3PO3 Pass Test (limit about 0.1%)
PO3 0.001%
Sulfur compounds (as S) 0.002%
Heavy Metals (as Pb) 5 ppm
Fe 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty