Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tannic acid ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tannin

36410-22 036410-22 36410-36 036410-36
AA36410-36EA 192.98 USD InStock
AA36410-36 CAAA36410-36 AA36410-22 CAAA36410-22
Tannic acid ACS
Tannic acid


Formula: C76H52O46
MW: 1701.2 g/mol
Melting Pt: ∼ 218 °C (dec.)
Density: >1 g/cm³ (20 °C)
Flash Pt: 199 °C (390 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221
Order Now
Specification Test Results
Identification Pass test
Loss on drying 0.12
Residue after ignition 0.5%
Heavy Metals (as Pb) 0.003%
Zn 0.005%
Sugars & dextrin Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty