Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydriodic acid 55 - 58%, liquid ACS for use as a catalyst

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen iodideHydrogen iodide solutionHydroiodic acid

036484-AD 036484-AP 036484-AY
AA36484-AYEA 1085.99 USD InStock
AA36484-AY AA36484-AP AA36484-AD
Hydriodic acid 55 - 58%, liquid ACS for use as a catalyst
Hydriodic acid
Suitable for use in methoxyl determinations.

Used as catalyst for the conversion of primary alcohol to alkyl halide.

Caution: Light sensitive liquid.


Formula: HI
MW: 127.91 g/mol
Boiling Pt: 127 °C (1013 hPa)
Density: 1.701
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,II
Order Now
Specification Test Results
Assay (Unspecified) 55.0 - 58.0%
Residue on Ignition Max. 0.01%
Impurity Content, Chloride and Bromide (as Cl) Max. 0.05%
Impurity Content, Fe Max. 0.001%
Impurity Content, I₂ Max. 0.75%
Impurity Content, PO₄ Max. 0.001%
Impurity Content, SO₄ Max. 0.005%
Trace Metals (as Pb) Max. 0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty