Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formic acid ≥88% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methanoic acid 88%

36504-AP 036504-AP 036504-K3 36504-K3 36504-AY 036504-AY
AA36504-K3EA 149.11 USD InStock
AA36504-K3 AA36504-AY CAAA36504-K3 CAAA36504-AY AA36504-AP CAAA36504-AP
Formic acid ≥88% ACS
Formic acid


Formula: CH₂O₂
MW: 46.03 g/mol
Boiling Pt: 100…101 °C
Melting Pt: 7…9 °C
Density: 1.220
Flash Pt: 50 °C (156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
Merck Index: 13,04265
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/23/723723.jpg
Order Now
Specification Test Results
Color (APHA) 15
Dilution test Pass Test
Evaporation residue 0.002
CH3COOH 0.004
NH3 0.00005
Cl 0.001%
SO3 0.002% max.
SO3 Pass test
Heavy Metals (as Pb) 5 ppm
Fe 5ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty