Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pararosaniline hydrochloride 90+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride Basic Red 9 Pararosaniline chloride

36540-18 36540-30 036540-18 36540-30
AA36540-18EA 142.99 USD InStock
AA36540-18 CAAA36540-18 AA36540-30 CAAA36540-30
Pararosaniline hydrochloride 90+%
Pararosaniline hydrochloride
pH indicator 1.2 (purple) to 3.0 (red)


Formula: C₁₉H₁₈ClN₃
MW: 323.82 g/mol
Melting Pt: 268…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001657
CAS Number: 569-61-9
EINECS: 209-321-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty