Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formic acid ≥96% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methanoic acid 97%

36617-36 036617-A3 36617-57 036617-36 36617-A3 36617-57
AA36617-A3EA 198.99 USD InStock
AA36617-A3 AAA36617-57 AA36617-36 CAAA36617-36 CAAA36617-A3 CAAA36617-57
Formic acid ≥96% ACS
Formic acid


Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C
Melting Pt: 7…9 °C
Density: 1.220
Flash Pt: 50 °C (156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/23/723723.jpg
Order Now
Specification Test Results
Form Liquid
Dilution test Pass test
Evaporation residue 0.003%
CH3COOH 0.4%
NH3 0.005%
Cl 0.001%
SO3 0.003%
SO3 Pass test
Heavy Metals (as Pb) 0.001%
Fe 0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty