Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloroethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

36636-K2 36636-AK 036636-K2 036636.AD 036636-AK 36636-AD
AA36636-K2EA 284.25 USD InStock
CAAA36636-AK AA36636-K2 CAAA36636-K2 AAA36636-AD CAAAA36636-AD AA36636-AK
2-Chloroethanol 99%
2-Chloroethanol


Formula: C₂H₅ClO
MW: 80.51 g/mol
Boiling Pt: 129 °C
Melting Pt: –63 °C
Density: 1.202
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002829
CAS Number: 107-07-3
EINECS: 203-459-7
UN: 1135
ADR: 6.1,I
Merck Index: 13,03827
https://us-legacy.vwr.com/stibo/web/std.lang.all/79/86/727986.jpg
Order Now
Specification Test Results
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty