Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) telluride ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Mercury telluride Mercurous telluride

036666-04 36666-09 036666-09 36666-04
AA36666-04EA 132.99 USD InStock
AA36666-04 CAAA36666-04 CAAA36666-09 AA36666-09
Mercury(II) telluride ≥99.999% (metals basis)
Mercury (II) telluride


Formula: HgTe
MW: 328.19 g/mol
Melting Pt: 673 °C
Density: 8.63
Storage Temperature: Ambient
MDL Number: MFCD00016133
CAS Number: 12068-90-5
EINECS: 235-108-9
UN: 2025
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty