Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl red sodium salt ACS

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt Sodium 2-(p-(dimethylamino)phenylazo)benzoate Methyl red water soluble

036667-14 36667-14 036667-22 36667-22
AA36667-22EA 147.2 USD InStock
CAAA36667-14 AA36667-22 AA36667-14 CAAA36667-22
Methyl red sodium salt ACS
Methyl red sodium salt


Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Melting Pt: > 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142
Order Now
Specification Test Results
Clarity of alcohol solution Pass test
Clarity of aqueous solution Pass test
Visual transition interval from pH 4.2 (pink) to 6.2 (yellow)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty