Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Starch, soluble modified (insolubles 0.01% max)

Supplier: Thermo Scientific Chemicals
Synonyms: Amylodextrin Vitex® iodine indicator

36673-36 36673-22 036673-22 036673-36
AA36673-36EA 168.71 USD InStock
AA36673-36 AA36673-22 CAAA36673-36 CAAA36673-22
Starch, soluble modified (insolubles 0.01% max)
Starch, soluble
For iodometric titrations

Instantly soluble in water


Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty