Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)benzaldehyde ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(N,N-Dimethylamino)benzaldehyde

36684-36 36684-22 036684-36 036684-14 36684-14 036684-22
AA36684-36EA 446.94 USD InStock
AA36684-36 AA36684-14 CAAA36684-22 CAAA36684-36 CAAA36684-14 AA36684-22
4-(Dimethylamino)benzaldehyde ACS
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (17 mmHg)
Melting Pt: 73…75 °C
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: 164 °C (327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/54/746254.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty