Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: CDTA monohydrate CyDTA monohydrate trans-Cyclohex-1,2-ylenediaminetetra(acetic acid) monohydrate trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate

36697-06 36697-14 036697-06 36697-22 036697-36 036697-14 036697-22
AA36697-06EA 24.99 USD InStock
CAAA36697-06 AA36697-06 AAA36697-36 AA36697-22 CAAA36697-14 AA36697-14 CAAA36697-22
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate


Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9
Order Now
Specification Test Results
Form Solid
Heavy Metals (as Pb) 0.001%
Fe 0.005%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty