Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


038292-09 038292-18 38292-09 38292-18
AA38292-18EA 360.99 USD InStock
AA38292-18 CAAA38292-09 CAAA38292-18 AA38292-09
Palladium 1% on activated carbon powder wetted with ca. 50% water, reduced
Palladium
Sold on a dry weight basis. Unit weight excludes water weight

Physical Form: 900-1100 m²/g


Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS Number: 7440-05-3
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty