Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

38367-22 038367-22 038367-36 38367-36
AA38367-22EA 52.34 USD InStock
CAAA38367-22 AA38367-22 CAAA38367-36 AA38367-36
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Cerium(III) ammonium nitrate tetrahydrate


Formula: H₈CeN₇O₁₅.4H₂O
MW: 558.28 g/mol (486.22 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00210608
CAS Number: 13083-04-0
UN: 1477
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty