Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) 2-ethylhexanoate 40% (w/w) in mineral spirits (6% Mn)

Supplier: Thermo Scientific Chemicals
Danger

38607-A7 38607-30 038607-30 38607-A7
AA38607-30EA 28.89 USD InStock
AA38607-30 CAAA38607-30 AA38607-A7 CAAA38607-A7
Manganese(II) 2-ethylhexanoate 40% (w/w) in mineral spirits (6% Mn)
Manganese(II) 2-ethylhexanoate


Formula: C₁₆H₃₀MnO₄
Density: 0.897
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00058698
CAS Number: 13434-24-7
UN: 1268
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty