Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl orange 0.1% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Synonyms: C.I. acid orange 52p-[[p-(dimethylamino)phenyl]azo]benzenesulfonic acid sodium salt

38695-AP 038695-AP
AA38695-APEA 48.36 USD InStock
CAAA38695-AP AA38695-AP
Methyl orange 0.1% (w/v) in aqueous solution
Methyl orange


Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Density: 0.987
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty