Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Petroleum spirit, 35…60 °C for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Petroleum ether

038985-K2 38985-K7 038985-M1 38985-M1 38985-K2 038985-K7
AA38985-M1EA 190.77 USD InStock
AA38985-M1 CAAA38985-K2 AA38985-K2 CAAA38985-K7 AA38985-K7 CAAA38985-M1
Petroleum spirit, 35…60 °C for HPLC
Petroleum spirit


Boiling Pt: 35…60 °C
Density: 0.640
Flash Pt: –49 °C (–57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty