Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: M-PyrolN-Methyl-2-pyrrolidinoneN-Methyl-2-pyrrolidoneN-Methyl-alpha-pyrrolidoneN-Methyl-gamma-butyrolactamN-MethylpyrrolidinoneN-methylpyrrolidoneN-MethylpyrrolidoneNMP

038986-K7 38986-K2 38986-M1 038986-K2 038986-M1 38986-K7
AA38986-K2EA 219.19 USD InStock
CAAA38986-K2 AA38986-K2 CAAA38986-K7 AA38986-K7 CAAA38986-M1 AA38986-M1
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC
N-Methyl-2-pyrrolidone (NMP)


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 204 °C
Melting Pt: –24 °C
Density: 1.033
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty