Would you like to log in again?
Beakers, Copper, Thermo Scientific
Supplier: Thermo Scientific
039052-KT 39054-KT 039056-KT 039053-KT
AA39052-KTEA 136.99 USD InStock
AA39052-KT AA39056-KT AA39054-KT AA39053-KT
Beakers, Copper, Thermo Scientific
Beakers
These beakers are made up of copper.
  • Provides high electrical and thermal conductivities
  • High corrosive resistance
  • Low toxicity
Copper is considered as one of the most important metallic elements, due to its unique physical and chemical properties. With similar reactivity to silver and gold, it is classified as a noble metal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty