Would you like to log in again?
Aluminium Dishes, with Flat bottom and Straight Sides, Thermo Scientific
Supplier: Thermo Scientific
39069-UD 39073-KH 39069-KH 39077-UD 39077-KH 39083-KH 39074-UD 39084-KH 39079-KH 39074-KH 39073-UD 39079-UD 039072-UD 39084-UD 39076-KH 39080-KH 39080-UD 039072-KH
AA39080-UDEA 68.02 USD InStock
AA39080-UD AA39073-KH AA39072-KH AA39076-KH AA39084-UD AA39069-UD AA39077-KH AA39074-KH AA39079-KH AA39080-KH AA39074-UD AA39073-UD AA39072-UD AA39069-KH AA39077-UD AA39083-KH AA39084-KH AA39079-UD
Aluminium Dishes, with Flat bottom and Straight Sides, Thermo Scientific
Trays/Dishes/Pots Evaporating Basins/Dishes
These beakers are made up of aluminium
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Designed to provide long life, increase efficiency and lower the running cost.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty